Learn More
PFEIFFER VACUUM INC Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 22, Ultimate Vacuum: 3E-3mbar, Pumping Speed: 11.5 CFM
Catalog No. NC3978943
Encompass_Preferred
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 22, Ultimate Vacuum: 3E-3mbar, Pumping Speed: 11.5CFM, Noise Level: 52 dB(A), Weight: 32kg, Dimensions (L x W x H): 483 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF
Catalog No. NC3978943 Supplier PFEIFFER VACUUM INC Supplier No. 2000239230
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 22, Ultimate Vacuum: 3E-3mbar, Pumping Speed: 11.5CFM, Noise Level: 52 dB(A), Weight: 32kg, Dimensions (L x W x H): 483 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF