Learn More
PFEIFFER VACUUM INC Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 6, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 3.8CFM
Catalog No. NC3978931
Encompass_Preferred
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 6, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 3.8CFM, Noise Level: 50 dB(A), Weight: 25.5kg, Dimensions (L x W x H): 413 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF
Catalog No. NC3978931 Supplier PFEIFFER VACUUM INC Supplier No. 2000239246
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 6, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 3.8CFM, Noise Level: 50 dB(A), Weight: 25.5kg, Dimensions (L x W x H): 413 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF