Learn More
Cole-Parmer Luer to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Male Luer; 10/Pk
Catalog No. 502558191 Shop All Cole-Parmer Products
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
Catalog No. 50-255-8191 Supplier Cole-Parmer Supplier No. 5011036
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.