Learn More
THORLABS INC Rotating Clamp for Ø1/2" Posts, 360° Continuously Adjustable, 5 mm Hex
Catalog No. NC1118096
Encompass
Rotating Clamp for Ø1/2" Posts, 360° Continuously Adjustable, 5 mm Hex
Catalog No. NC1118096 Supplier THORLABS INC Supplier No. SWC/M
Rotating Clamp for Ø1/2" Posts, 360° Continuously Adjustable, 5 mm Hex