Learn More
Thomas Scientific Denviller Benchtop Storage, Rotating Wash Bottle Holder (8.5W x 8.5D x 9H)
SDP
Catalog No. 501460006
Encompass_Preferred
Rotating Wash Bottle Holder (H0382): Stay mobile with the Rotating Wash Bottle Holder; The sturdy handle provides comfort and control; Holds bottles up to 3” in diameter; Polycarbonate construction Rotating Two-Tier Lab Depot (H0381): Holds up to four wash bottles; Rotates 360° degrees; Store all your laboratory essentials in the four large open compartments; Includes two removable wash bottle inserts; Made of ABS for easy disinfecting with bleach solution Quad Bottle Holder with Storage (H0226): Smaller footprint increases work space; Holds up to four wash bottles; Made of ABS for easy disinfecting with bleach solution; Can be wall mounted Centrifuge Balance Tube Caddy (C0021): 17mm hole diameter; Saves bench space and keeps tubes within reach; Special adhesive-mount tape comes pre-attached to caddy and withstands centrifuge vibration
Catalog No. 50-146-0006 Supplier Thomas Scientific Supplier No. 1159R61
Rotating Wash Bottle Holder (H0382): Stay mobile with the Rotating Wash Bottle Holder; The sturdy handle provides comfort and control; Holds bottles up to 3” in diameter; Polycarbonate construction Rotating Two-Tier Lab Depot (H0381): Holds up to four wash bottles; Rotates 360° degrees; Store all your laboratory essentials in the four large open compartments; Includes two removable wash bottle inserts; Made of ABS for easy disinfecting with bleach solution Quad Bottle Holder with Storage (H0226): Smaller footprint increases work space; Holds up to four wash bottles; Made of ABS for easy disinfecting with bleach solution; Can be wall mounted Centrifuge Balance Tube Caddy (C0021): 17mm hole diameter; Saves bench space and keeps tubes within reach; Special adhesive-mount tape comes pre-attached to caddy and withstands centrifuge vibration