Learn More
Electron Microscopy Sciences Rotamix™ Rotation Rod Tt1.5
SDP
Catalog No. 50996366
Encompass_Preferred
Rod for 34 - 1.5 ml tubes (9 – 12mm).
Catalog No. 50-996-366 Supplier Electron Microscopy Sciences Supplier No. 6106210
Rod for 34 - 1.5 ml tubes (9 – 12mm).