Learn More
Chemglass Life Sciences Rotator Genie, Magnetic Platform, 120V
SDP
Catalog No. 50143397
Encompass
Rotator Genie, Magnetic Platform, 120V. The Rotator Genie™ holds a wide variety of tubes, flasks, microplates, bags and odd-shaped vessels at any angle. Available in two interchangeable platform configurations. Standard platform is designed to turn 90°, allowing different mixing options. The magnetic platform features unique spring clip plate system. Designed for cold room and incubator use and will easily fit in most incubators. Uniquely designed for a wide selection of motions which can be adjusted from gentle rolling to forceful, end over end mixing. Continuous-duty quality motor provides smooth, quiet operation at 5 to 35RPM.
Catalog No. 50-143-397 Supplier Chemglass Life Sciences Supplier No. CLS3962003
Rotator Genie, Magnetic Platform, 120V. The Rotator Genie™ holds a wide variety of tubes, flasks, microplates, bags and odd-shaped vessels at any angle. Available in two interchangeable platform configurations. Standard platform is designed to turn 90°, allowing different mixing options. The magnetic platform features unique spring clip plate system. Designed for cold room and incubator use and will easily fit in most incubators. Uniquely designed for a wide selection of motions which can be adjusted from gentle rolling to forceful, end over end mixing. Continuous-duty quality motor provides smooth, quiet operation at 5 to 35RPM.