Learn More
Research Products International Corp Rotator Genie w/ Magnetic Platform
SDP
Catalog No. 501976951
Encompass
Designed to provide variable speed rotating in a compact, durable and user-friendly unit. The units compact footprint will fit into most incubators allowing for controlled environment mixing. Has a durable thermoplastic platform that securely holds vessels with super stretch latex rubber strapping. Ideal for mixing of almost any kind of laboratory samples. Supplied with magnetic platform.
Catalog No. 50-197-6951 Supplier Research Products International Corp Supplier No. 155593
Designed to provide variable speed rotating in a compact, durable and user-friendly unit. The units compact footprint will fit into most incubators allowing for controlled environment mixing. Has a durable thermoplastic platform that securely holds vessels with super stretch latex rubber strapping. Ideal for mixing of almost any kind of laboratory samples. Supplied with magnetic platform.