Learn More
Research Products International Corp Rotator Genie w/ Standard Platform
SDP
Catalog No. 501976955
Encompass
Designed to provide variable speed rotating in a compact, durable and user-friendly unit. The units compact footprint will fit into most incubators allowing for controlled environment mixing. Has a durable thermoplastic platform that securely holds vessels with super stretch latex rubber strapping. Ideal for mixing of almost any kind of laboratory samples. Supplied with platform. Continuous duty motor provides smooth quiet operation in a wide temperature range of 0° to 38°C.
Catalog No. 50-197-6955 Supplier Research Products International Corp Supplier No. 155592
Designed to provide variable speed rotating in a compact, durable and user-friendly unit. The units compact footprint will fit into most incubators allowing for controlled environment mixing. Has a durable thermoplastic platform that securely holds vessels with super stretch latex rubber strapping. Ideal for mixing of almost any kind of laboratory samples. Supplied with platform. Continuous duty motor provides smooth quiet operation in a wide temperature range of 0° to 38°C.