Learn More
Electron Microscopy Sciences Rotatrim Rolling Trimmer Side Guide
SDP
Catalog No. 5028397
Encompass_Preferred
Mounts on the squaring arm of the cutter then slides to the desired position and locks in place. It is used when cutting sheet materials to identical lengths and can be used with either the Dualrail or the Monorail RotaTrim Cutters.
Catalog No. 50-283-97 Supplier Electron Microscopy Sciences Supplier No. 7488210
Mounts on the squaring arm of the cutter then slides to the desired position and locks in place. It is used when cutting sheet materials to identical lengths and can be used with either the Dualrail or the Monorail RotaTrim Cutters.