Learn More
BUCHI Rotavapor R-100 Vertical Condenser Glass Assembly
SDP
Catalog No. 05405048 Shop All BUCHI Products
Change view
Click to view available options
No Safety Coating
Catalog No.
05-405-048 No Safety Coating
1 options
Catalog No. 05-405-048 Supplier BUCHI Supplier No. 11057056
Add to Cart
Add to Cart
Vertical Condenser Glass Assembly for Rotavapor R-100, 1500 cm2. For standard distillations, used with recirculating chiller or tap water. Includes a 1 liter receiving flask, required tubing, and a ball joint clamp.
  • Includes receiving flask (1 L), tubing, and a ball joint clamp
  • Does NOT include an evaporating flask, vacuum seal, vapor duct, or condenser holder
  • For use with Rotavapor R-100, R-3
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condensor
For Use With (Equipment) Rotavapor R-100, R-3
Includes Receiving Flask (1 L), Tubing, and a Ball Joint Clamp
Material (Coating) Uncoated
Volume (Metric) 1000 mL