Learn More
BUCHI Rotavapor R-300
GSA_VASDP
Catalog No. 05001055 Shop All BUCHI Products
Change view
Click to view available options
Condenser Type:
Cold Trap
Vertical Coil
Temperature Range (Metric):
20°C to 220°C
Includes:
I-300 Interface, V-300 Pump, Woulff Bottle
Woulff Bottle
Model:
Electronic Lift System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Condenser Type Temperature Range (Metric) Includes Model
05-001-055 Cold Trap 20°C to 220°C I-300 Interface, V-300 Pump, Woulff Bottle Electronic Lift System
05-000-948 Cold Trap 20°C to 220°C Woulff Bottle Electronic Lift System
05-000-978 Vertical Coil 20°C to 220°C Woulff Bottle Electronic Lift System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-001-055 Supplier BUCHI Supplier No. 11SR300251CP02
Add to Cart
Add to Cart
Request a Quote
Buchi Rotavapor R-300 is a convenient and versatile rotary evaporator.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Voltage 100/120 V
Capacity (Metric) 5 L
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 220°C
Material (Coating) Safety Coating (P+G)
Includes I-300 Interface, V-300 Pump, Woulff Bottle
Condenser Type Cold Trap
Glassware Assembly Type Cold trap (C)
Type Rotary Evaporator
Model Electronic Lift System
Standard Taper Joint SJ 24/40
Timer Yes