Learn More
BUCHI Rotavapor R-300 Splash Shield
SDP
Catalog No. 05405075 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-300
Catalog No. For Use With (Equipment)
05-405-075 Rotavapor R-300
1 options
Catalog No. 05-405-075 Supplier BUCHI Supplier No. 11064768
Add to Cart
Add to Cart
Splash shield is fixed onto Rotavapor R-300 with the opening on the front side, only one hand needed for easy handling.
  • Opening on the front side for easy handling
  • Constructed of 6 mm thick polycarbonate
  • For use with Rotavapor R-300
Specifications
Description Thickness: 6 mm
Material Polycarbonate
Type Splash Shield
For Use With (Application) Splash Protection
For Use With (Equipment) Rotavapor R-300