Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
SDP
Catalog No. 05405063 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Application):
Glass Assembly A
Glass Assembly V, C, S, E, CR, BY, HP
KD22
Standard Taper Joint Number Center:
24/40
29/32
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Standard Taper Joint Number Center
05-405-063 Glass Assembly A 24/40
05-405-062 Glass Assembly V, C, S, E, CR, BY, HP 24/40
05-405-022 KD22 29/32
05-405-061 Glass Assembly V, C, S, E, CR, BY, HP 29/32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05-405-063 Supplier BUCHI Supplier No. 11062268
Add to Cart
Add to Cart
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
  • Combi-Clip is included for fastening the evaporating flask
Specifications
Material Borosilicate Glass (3.3)
Type Vapor Duct
For Use With (Application) Glass Assembly A
For Use With (Equipment) Rotavapor R-300
Includes Combi-Clip
Standard Taper Joint Number Center 24/40