Learn More
ACROBiosystems Rotavirus A VP4 Protein, His Tag (MALS verified)
Catalog No. 502330951
Encompass_Preferred
Rotavirus A (strain RVA/Human/United States/Wa/1974/G1P1A[8]) VP4 Protein, His Tag (VP4-R5243) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Asn 476 (Accession # P11193).
Catalog No. 50-233-0951 Supplier ACROBiosystems Supplier No. VP4R5243100UG
Rotavirus A (strain RVA/Human/United States/Wa/1974/G1P1A[8]) VP4 Protein, His Tag (VP4-R5243) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Asn 476 (Accession # P11193).