Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Rotavirus Rapid Test Control Pack
SDP

Catalog No. 22158094 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-094 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-094 Supplier ZeptoMetrix Supplier No. KZMC036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for in vitro rapid tests determination of presence or absence of Rotavirus. ZeptoMetrix Rotavirus Rapid Test Control Pack is designed for monitoring assay performance.

  • Composed of two members representing Rotavirus positive and negative samples
  • Panel member contains 1.5mL of artificial nasal matrix and inactivated Rotavirus
  • Used for training, lot-to-lot comparison of reagent test kits and to evaluate and compare intra and inter laboratory performance
  • Prepared from human and non-human components
  • Inactivated to reduce infectious risk

Reagents:

  • One vial Rotavirus (WA) Positive (1.5mL)
  • One vial Rotavirus Negative (1.5mL)

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Rapid Test Control Pack
For Use With (Application) In vitro rapid tests for the determination of the presence or absence of Rotavirus
Quantity 1.5 mL

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.