Learn More
Enzo Life Sciences Rotenone, (1g), CAS Number: 83-79-4
SDP
Catalog No. 501031745
Encompass_Preferred
Insecticide. Inhibitor of mitochondrial electron transport. Specifically inhibits NAD-linked substrate oxidation at the oxygen side of NADH dehydrogenase (EC50=10pmol/mg). Inhibits mammalian cell proliferation by inhibiting microtubule assembly through tubulin binding. Can inhibit mitochondrial swelling and cytochrome c release induced by N-methyl-4-phenylpyridinium. Formula: C23H22O6. MW: 394.4. CAS Number: 83-79-4. MI: 14: 8271. Purity: ≥97% (UHPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C
Catalog No. 50-103-1745 Supplier Enzo Life Sciences Supplier No. ALX350360G001
Insecticide. Inhibitor of mitochondrial electron transport. Specifically inhibits NAD-linked substrate oxidation at the oxygen side of NADH dehydrogenase (EC50=10pmol/mg). Inhibits mammalian cell proliferation by inhibiting microtubule assembly through tubulin binding. Can inhibit mitochondrial swelling and cytochrome c release induced by N-methyl-4-phenylpyridinium. Formula: C23H22O6. MW: 394.4. CAS Number: 83-79-4. MI: 14: 8271. Purity: ≥97% (UHPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C