Learn More
Medchemexpress LLC HY-B1756 5g , Rotenone CAS:83-79-4 Purity:98%
SDP
Catalog No. 501492218
Encompass_Preferred
Medchemexpress, HY-B1756 5g Rotenone CAS:83-79-4 Formula:C23H22O6 Purity:98% Rotenone is an inhibitor of mitochondrial electron transport at NADH:ubiquinone oxidoreductase, and is used to induce a Parkinson-like syndrome as an experimental model in rats. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2218 Supplier Medchemexpress LLC Supplier No. HYB17565G
Medchemexpress, HY-B1756 5g Rotenone CAS:83-79-4 Formula:C23H22O6 Purity:98% Rotenone is an inhibitor of mitochondrial electron transport at NADH:ubiquinone oxidoreductase, and is used to induce a Parkinson-like syndrome as an experimental model in rats. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.