Learn More
Medchemexpress LLC HY-B1756 500mg , Rotenone CAS:83-79-4 Purity:>98%
SDP
Catalog No. 502259117
Encompass_Preferred
HY-B1756 500mg Rotenone CAS: 83-79-4 Rotenone is an mitochondrial electron transport chain complex I inhibitor. Rotenone induces apoptosis through enhancing mitochondrial reactive oxygen species production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9117 Supplier Medchemexpress LLC Supplier No. HYB1756500MG
HY-B1756 500mg Rotenone CAS: 83-79-4 Rotenone is an mitochondrial electron transport chain complex I inhibitor. Rotenone induces apoptosis through enhancing mitochondrial reactive oxygen species production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.