Learn More
Enzo Life Sciences Rotenone (5g). CAS: 83-79-4
SDP
Supplier:  Enzo Life Sciences ALX350360G005
Encompass_Preferred
Insecticide. Inhibitor of mitochondrial electron transport. Specifically inhibits NAD-linked substrate oxidation at the oxygen side of NADH dehydrogenase (EC50=10pmol/mg). Inhibits mammalian cell proliferation by inhibiting microtubule assembly through tubulin binding. Can inhibit mitochondrial swelling and cytochrome c release induced by N-methyl-4-phenylpyridinium. Purity: ≥97% (UHPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8860