Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotenone 97%, Thermo Scientific™

Catalog No. AC132370250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC132370250 25 g
AC132370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132370250 Supplier Thermo Scientific Chemicals Supplier No. 132370250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-79-4
Molecular Formula C23H22O6
Molecular Weight (g/mol) 394.423
InChI Key JUVIOZPCNVVQFO-HBGVWJBISA-N
Synonym rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
PubChem CID 6758
ChEBI CHEBI:28201
SMILES CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC

Specifications

Melting Point 156°C to 163°C
Color Yellow to Beige
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 19, II, 438
Merck Index 15, 8403
Formula Weight 394.41
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Rotenone, 97%
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-501-9
RTECSNumber DJ2800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.