Learn More
Abcam Rotenone, mitochondrial electron transport chain inhibitor, 5G
Catalog No. 502706130
Encompass_Preferred
MW 394.4 Da, Purity >97%. Mitochondrial electron transport chain complex I inhibitor (IC₅₀ values are 1.7 - 2.2 μM). Potent NADH oxidation inhibitor (IC₅₀ = 3.4 nM). Suppresses microtubule assembly. Induces BRL cell apoptosis. Shows antiproliferative effects. Shows central effects. Induces parkinsonian behavior in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6130 Supplier Abcam Supplier No. AB1431455G
MW 394.4 Da, Purity >97%. Mitochondrial electron transport chain complex I inhibitor (IC₅₀ values are 1.7 - 2.2 μM). Potent NADH oxidation inhibitor (IC₅₀ = 3.4 nM). Suppresses microtubule assembly. Induces BRL cell apoptosis. Shows antiproliferative effects. Shows central effects. Induces parkinsonian behavior in vivo. .
The product is subject to the following: Abcam Restricted Use Statement