Learn More
Medchemexpress LLC Rotenone (Standard) | 83-79-4 | 99.8% | 250 MG
SDP
Catalog No. 5000414433
Encompass_Preferred
Rotenone (Standard) is the analytical standard of Rotenone. It is intended for research and analytical applications. It functions as a mitochondrial electron transport chain complex I inhibitor and induces apoptosis by enhancing mitochondrial reactive oxygen species production.
  • Intended for research and analytical applications
  • Functions as a mitochondrial electron transport chain complex I inhibitor
  • Induces apoptosis by enhancing mitochondrial reactive oxygen species production
  • Used as a reference standard supplied assay
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-14433 Supplier Medchemexpress LLC Supplier No. 250MGHYB1756R
Rotenone (Standard) is the analytical standard of Rotenone. It is intended for research and analytical applications. It functions as a mitochondrial electron transport chain complex I inhibitor and induces apoptosis by enhancing mitochondrial reactive oxygen species production.
  • Intended for research and analytical applications
  • Functions as a mitochondrial electron transport chain complex I inhibitor
  • Induces apoptosis by enhancing mitochondrial reactive oxygen species production
  • Used as a reference standard supplied assay
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS