Learn More
Medchemexpress LLC Rotigaptide 1mg | 355151-12-1 | 1MG
SDP
Catalog No. 5000243663
Encompass_Preferred
Rotigaptide (ZP123) is a novel and specific modulator of connexin 43 (Cx43) Rotigaptide prevents the uncoupling of Cx43-mediated gap junction communication and normalizes cell-to-cell communication during acute metabolic stress Rotigaptide is a potent antiarrhythmic peptide (AAP) with improved stability and has the potential for the investigation of cardiac arrhythmias-specifically atrial fibrillation[1 [2
Catalog No. 50-002-43663 Supplier Medchemexpress LLC Supplier No. HY1062251MG
Rotigaptide (ZP123) is a novel and specific modulator of connexin 43 (Cx43) Rotigaptide prevents the uncoupling of Cx43-mediated gap junction communication and normalizes cell-to-cell communication during acute metabolic stress Rotigaptide is a potent antiarrhythmic peptide (AAP) with improved stability and has the potential for the investigation of cardiac arrhythmias-specifically atrial fibrillation[1 [2