Learn More
Medchemexpress LLC Rotigotine Hydrochloride | 125572-93-2 | 99.45% | C19H26ClNOS | 500 MG
SDP
Catalog No. 5000433335
Encompass_Preferred
Rotigotine Hydrochloride is a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It is suitable for research use only.
  • Full agonist of dopamine receptor
  • Partial agonist of the 5-HT1A receptor
  • Antagonist of the α2B-adrenergic receptor
  • Ki values of 0.71 nM for the dopamine D3 receptor
  • Ki values of 4-15 nM for D2, D5, and D4 receptors
  • Ki value of 83 nM for the dopamine D1 receptor
  • White to off-white solid appearance
  • Suitable for research use only
Catalog No. 50-004-33335 Supplier Medchemexpress LLC Supplier No. HYA0007500MG
Rotigotine Hydrochloride is a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It is suitable for research use only.
  • Full agonist of dopamine receptor
  • Partial agonist of the 5-HT1A receptor
  • Antagonist of the α2B-adrenergic receptor
  • Ki values of 0.71 nM for the dopamine D3 receptor
  • Ki values of 4-15 nM for D2, D5, and D4 receptors
  • Ki value of 83 nM for the dopamine D1 receptor
  • White to off-white solid appearance
  • Suitable for research use only