Learn More
Medchemexpress LLC Rotigotine Hydrochloride | 125572-93-2 | 99.5% | C19H26ClNOS | 1 ML
SDP
Catalog No. 5000433498
Encompass_Preferred
Rotigotine Hydrochloride (N-0923 Hydrochloride) is a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It has a Ki of 0.71 nM for the dopamine D3 receptor, 4-15 nM for the D2, D5, and D4 receptors, and 83 nM for the dopamine D1 receptor. It is intended for research use only.
  • Full agonist of dopamine receptor
  • Partial agonist of 5-HT1A receptor
  • Antagonist of α2B-adrenergic receptor
  • Ki of 0.71 nM for dopamine D3 receptor
  • Ki of 4-15 nM for D2, D5, and D4 receptors
  • Ki of 83 nM for dopamine D1 receptor
  • For research use only
Catalog No. 50-004-33498 Supplier Medchemexpress LLC Supplier No. HYA000710MM/1ML
Rotigotine Hydrochloride (N-0923 Hydrochloride) is a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It has a Ki of 0.71 nM for the dopamine D3 receptor, 4-15 nM for the D2, D5, and D4 receptors, and 83 nM for the dopamine D1 receptor. It is intended for research use only.
  • Full agonist of dopamine receptor
  • Partial agonist of 5-HT1A receptor
  • Antagonist of α2B-adrenergic receptor
  • Ki of 0.71 nM for dopamine D3 receptor
  • Ki of 4-15 nM for D2, D5, and D4 receptors
  • Ki of 83 nM for dopamine D1 receptor
  • For research use only