Learn More
Hettich Centrifuges ROTINA 380, (4) 250/290mL,Package
SDP
Supplier:  Hettich Centrifuges 380BOTTLE
Encompass_Preferred
Hettich ROTINA 380 BOTTLE PACKAGE, (1) Centrifuge w/Temp. Control, (1) Horizontal Rotor, (4) Buckets, (4) Adapters, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7617
Add to Cart
Request a Quote