Learn More
Hettich Centrifuges ROTINA 420,(4)400 mL,Bio,Package
SDP
Catalog No. 502537596 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 420 BOTTLE PACKAGE 400 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 400 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7596 Supplier Hettich Centrifuges Supplier No. 420BOTTLE400BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 420 BOTTLE PACKAGE 400 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 400 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years