Learn More
Hettich Centrifuges ROTINA 420 S/O ROTOR BUCKET
SDP
Supplier:  Hettich Centrifuges 4780
Encompass_Preferred
  • Designed for seamless integration with specific centrifuge models.
  • Crafted from light metal for durability and lightweight handling.
  • Employs a single-hand clamp lock mechanism for secure handling of samples.
  • Compatible with Hettich 4784-A swing-out rotor.
  • Works with Hettich ROTINA 420 and ROTINA 420 R centrifuge models.
  • Biocontainment capabilities when paired with an optional lid.
  • TÜV certified aerosol-tightness when equipped with the lid.
  • Integrated handle for easy loading, unloading, and transport.
  • Facilitates efficient sealing and handling of samples.
  • Recommended ordering in multiples of 4 for convenience.
Catalog No. NC3244357