Learn More
Hettich Centrifuges ROTINA 420R,(4)250 mL,Bio,Package
SDP
Catalog No. 502537589 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 420R BOTTLE PACKAGE 250 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 250 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7589 Supplier Hettich Centrifuges Supplier No. 420RBOTTLE250BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 420R BOTTLE PACKAGE 250 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 250 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years