Learn More
Hettich Centrifuges ROTINA380R,(18)15,(8)50,BioPackage
SDP
Catalog No. 502537603 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 380R CELL CULTURE PACKAGE 2 BC, (1) Centrifuge w/Temp. Control, (1) Horizontal Rotor, (4) Buckets (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7603 Supplier Hettich Centrifuges Supplier No. 380RCELLCULTURE2BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 380R CELL CULTURE PACKAGE 2 BC, (1) Centrifuge w/Temp. Control, (1) Horizontal Rotor, (4) Buckets (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years