Learn More
Hettich Centrifuges ROTINA380R,(64)13,(52)16,BioPk
SDP
Supplier:  Hettich Centrifuges 380RBLOOD4BC
Encompass_Preferred
Hettich ROTINA 380R BLOOD TUBE PACKAGE 4 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7607
Add to Cart
Request a Quote