Learn More
Hettich Centrifuges ROTINA380R,(64)13,(52)16,Package
SDP
Catalog No. 502537608 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 380R BLOOD TUBE PACKAGE 4, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7608 Supplier Hettich Centrifuges Supplier No. 380RBLOOD4
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 380R BLOOD TUBE PACKAGE 4, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years