Learn More
Hettich Centrifuges ROTINA420,(52)15,(20)50,BioPackage
SDP
Catalog No. 502537593 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 420 CELL CULTURE PACKAGE 4 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (4) Adapters 15 mL, (4) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7593 Supplier Hettich Centrifuges Supplier No. 420CELLCULTURE4BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 420 CELL CULTURE PACKAGE 4 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (4) Adapters 15 mL, (4) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years