Learn More
Hettich Centrifuges ROTINA420R,(26)15(10)50,BioPackage
SDP
Catalog No. 502537586 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 420R CELL CULTURE PACKAGE 2 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7586 Supplier Hettich Centrifuges Supplier No. 420RCELLCULTURE2BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 420R CELL CULTURE PACKAGE 2 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years