Learn More
Hettich Centrifuges ROTINA420R,(52)13,(36)16,BioPk
SDP
Catalog No. 502537591 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTINA 420R BLOOD TUBE PACKAGE 2 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (2) Adapters 13 x 75/100, (2) Adapters 16 x 75/100, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7591 Supplier Hettich Centrifuges Supplier No. 420RBLOOD2BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTINA 420R BLOOD TUBE PACKAGE 2 BC, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets (2) Adapters 13 x 75/100, (2) Adapters 16 x 75/100, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years