Learn More
Anchor Scientific ROTO FLOAT 20FT CABLE SWITCH
Catalog No. NC3652725
Encompass
Roto-Float® Signal-Duty Float Switches w/ 20-ft Cable, SPST, Normally Closed (NC), Suspended (Internal Weight)
Catalog No. NC3652725 Supplier Anchor Scientific Supplier No. 47748
Roto-Float® Signal-Duty Float Switches w/ 20-ft Cable, SPST, Normally Closed (NC), Suspended (Internal Weight)