Learn More
Electron Microscopy Sciences Roto Shake Genie Rotator/Rocker 220 V
SDP
Catalog No. 501899652
Encompass_Preferred
Combination platform/tilt, shaker, rocker, and specialized rotator mixes tubes by rotating, shaking, rolling, or any combination of these mixing actions. Easy to remove, two-sided stainless steel magnetic platform easily converts from rotating to rocking with a simple mechanical adjustment.
Magnetic platform accommodates up to two 6" x 6" plastic bags per side, a 12" x 12" non-skid metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side to accommodate similar or different tube sizes for various mixing solutions. Ideal for use in cold rooms and incubators for complete and even soaking of membrane-based blot assays/washings. Easily cleaned or decontaminated. Maintains the set speed between 32 degF (0 degree C) and 100 degF (38 degree C). Features magnetic platform and clip plates. Suitable for use in cold rooms and incubators. 2 year warranty.
Catalog No. 50-189-9652 Supplier Electron Microscopy Sciences Supplier No. 7183420
Combination platform/tilt, shaker, rocker, and specialized rotator mixes tubes by rotating, shaking, rolling, or any combination of these mixing actions. Easy to remove, two-sided stainless steel magnetic platform easily converts from rotating to rocking with a simple mechanical adjustment.
Magnetic platform accommodates up to two 6" x 6" plastic bags per side, a 12" x 12" non-skid metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side to accommodate similar or different tube sizes for various mixing solutions. Ideal for use in cold rooms and incubators for complete and even soaking of membrane-based blot assays/washings. Easily cleaned or decontaminated. Maintains the set speed between 32 degF (0 degree C) and 100 degF (38 degree C). Features magnetic platform and clip plates. Suitable for use in cold rooms and incubators. 2 year warranty.