Learn More
Lab Depot Inc Benchmark Roto-Mini Plus Rotator 5 to 70 rpm
SDP
Catalog No. NC2874119
Encompass_Preferred
Benchmark’s new Roto-Mini series of rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, etc. the mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing.
Catalog No. NC2874119 Supplier Lab Depot Inc Supplier No. R2024
Benchmark’s new Roto-Mini series of rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, etc. the mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing.