Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Roto-Shake Genie

Catalog No. 01804420 Shop All OHAUS Products
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-804-420 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-804-420 Supplier OHAUS Supplier No. 83041387
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for rocking, rotating, and orbital shaking.

  • A stainless steel Magnetic Platform Supplied with 1 Clip Plate each for 10-13 mm Tubes, 15-17 mm Tubes and 28-30 mm Tubes.
  • 4 Magnetic Bag Mounting Strips and Non-skid Tray.
TRUSTED_SUSTAINABILITY

Specifications

Rotating Angle 360°
Speed Range 5 to 35 rpm
Voltage 120 V
Product Type Rotator
Model Roto-Shake Genie
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.