Learn More
Research Products International Corp Roto-Shake Genie Platform Rotator/Rocker
SDP
Catalog No. 501976893
Encompass
Combines rocking and rotating in one rugged and versatile design. Magnetic Stainless Steel Platform and Clip Plate System holds a variety of container at any angle. Expansion kits available for increased capacity and different containers. Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings.
Supplied with magnetic platform (Part No. SI-1320), 1 clip plate each for 10-13mm tubes, 15-17mm tubes, and 28-30mm tubes (Part Nos. SI-1123, SI-1124 and SI-1125, respectively), 4 magnetic bag mounting strips (Part No. 0M-1126-201) and non-skid tray (Part No. 0A-1100-070).
Catalog No. 50-197-6893 Supplier Research Products International Corp Supplier No. 248816
Combines rocking and rotating in one rugged and versatile design. Magnetic Stainless Steel Platform and Clip Plate System holds a variety of container at any angle. Expansion kits available for increased capacity and different containers. Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings.
Supplied with magnetic platform (Part No. SI-1320), 1 clip plate each for 10-13mm tubes, 15-17mm tubes, and 28-30mm tubes (Part Nos. SI-1123, SI-1124 and SI-1125, respectively), 4 magnetic bag mounting strips (Part No. 0M-1126-201) and non-skid tray (Part No. 0A-1100-070).