Learn More
Alkali Scientific Roto-Therm™ Incubated Tube Rotator with Fixed Speed, 11.8 X 8.5 X 9", 115V
SDP
Catalog No. 502389312
Encompass_Preferred
Roto-Therm™ Incubated Rotator with Fixed Speed provides a wide range of mixing capabilities. Suitable for an array of laboratory applications including re-suspension, protein binding, nucleic acid purification, histochemistry, hybridization etc, the mixer includes interchangeable tube holders for accommodating all common tube sizes from 0.5mL to 50mL. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. For applications requiring temperature control, the Roto-Therm™ Incubated Tube Rotator can be digitally programmed to maintain chamber temperatures of ambient +5°C to 60°C and include an “in-lab” calibration function for ensuring accurate temperature control. Specifications: Temperature: Ambient +5 to 60°C Speed: 24rpm Dimensions: 11.8 x 8.5 x 9in (29.5 x 21.6 x 22.9cm) Weight: 9 lbs (4kg)
Catalog No. 50-238-9312 Supplier Alkali Scientific Supplier No. RS2020
Roto-Therm™ Incubated Rotator with Fixed Speed provides a wide range of mixing capabilities. Suitable for an array of laboratory applications including re-suspension, protein binding, nucleic acid purification, histochemistry, hybridization etc, the mixer includes interchangeable tube holders for accommodating all common tube sizes from 0.5mL to 50mL. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. For applications requiring temperature control, the Roto-Therm™ Incubated Tube Rotator can be digitally programmed to maintain chamber temperatures of ambient +5°C to 60°C and include an “in-lab” calibration function for ensuring accurate temperature control. Specifications: Temperature: Ambient +5 to 60°C Speed: 24rpm Dimensions: 11.8 x 8.5 x 9in (29.5 x 21.6 x 22.9cm) Weight: 9 lbs (4kg)