Learn More
Research Products International Corp Roto-Therm Plus Incubated Rotator w/tube holders
SDP
Catalog No. 502229814
Encompass
Rotating mixer provides wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization, etc. The mixer includes interchangeable tube holders that accommodate common tube sizes from 0.5ml to 50ml. Tube holders direction can be adjusted from vertical (end over end) mixing to horizontal (rolling) style mixing.
Chambers can be programmed to maintain ambient +5°C to 60°C and feature “in-lab” calibration function to ensure accurate temperature control. Available with variable speed control and advanced shaking/ mixing modes: Mix (Oscillating), Pause, Rock. Rotator includes tube holders for common tube sizes 24 x 0.5ml; 11 x 1.5-2.0ml (2/pk); 5 x 5-7ml (2/pk);4 x 10-15ml (2/pk); and 6 x 50ml(1/pk). Optional tube holders available for PCR and plates.
Catalog No. 50-222-9814 Supplier Research Products International Corp Supplier No. 260131
Rotating mixer provides wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization, etc. The mixer includes interchangeable tube holders that accommodate common tube sizes from 0.5ml to 50ml. Tube holders direction can be adjusted from vertical (end over end) mixing to horizontal (rolling) style mixing.
Chambers can be programmed to maintain ambient +5°C to 60°C and feature “in-lab” calibration function to ensure accurate temperature control. Available with variable speed control and advanced shaking/ mixing modes: Mix (Oscillating), Pause, Rock. Rotator includes tube holders for common tube sizes 24 x 0.5ml; 11 x 1.5-2.0ml (2/pk); 5 x 5-7ml (2/pk);4 x 10-15ml (2/pk); and 6 x 50ml(1/pk). Optional tube holders available for PCR and plates.