Learn More
Chemglass Life Sciences Roto-Therm Plus, Incubated Rotator (Fixed Speed), with Tube Holders, 115V
SDP
Supplier:  Chemglass Life Sciences CLS3969020
Encompass
Roto-Therm series of incubated rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization etc, the mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. For applications requiring temperature control, the Roto-Therm series can be digitally programmed to maintain chamber temperatures of ambient +5°C to 60°C and include an “in-lab” calibration function for ensuring accurate temperature control. Temperature: Ambient +5 to 60°C. Speed: 24rpm. The CLS-3969-020 is supplied with the following tube holders:  CLS-3969-032,  CLS-3969-036,  CLS-3969-040,  CLS-3969-044.  
Catalog No. 50-197-9470