Learn More
Thomas Scientific Roto-Therm Plus Incubated Rotator (variable speed) with tube holders, 115V
SDP
Catalog No. NC1568446
Encompass
Benchmark’s Roto-Therm series of incubated rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization etc, the mixers include interchangeable tube holders for accomomodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. For applications requiring temperature control, the Roto-Therm series can be digitally programmed to maintain chamber temperatures of ambient +5°C to 60°C and include an “in-lab” calibration function for ensuring accurate temperature control. The Roto-Therm is also available in a “Plus” configuration, providing the following additional features: Variable speed: 5 to 70rpm “Mix” function: Rotating in combination w/ oscillations (increased mixing) “Pause&rdq
Catalog No. NC1568446 Supplier Thomas Scientific Supplier No. 1159P17
Benchmark’s Roto-Therm series of incubated rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization etc, the mixers include interchangeable tube holders for accomomodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. For applications requiring temperature control, the Roto-Therm series can be digitally programmed to maintain chamber temperatures of ambient +5°C to 60°C and include an “in-lab” calibration function for ensuring accurate temperature control. The Roto-Therm is also available in a “Plus” configuration, providing the following additional features: Variable speed: 5 to 70rpm “Mix” function: Rotating in combination w/ oscillations (increased mixing) “Pause&rdq