Learn More
Alkali Scientific Alkali Scientific™ RotoBot Mini R4040 Lab Rotator with Tube Holder Starter Set
SDP
Catalog No. 503029546
Encompass_Preferred
Advanced programming with a wide variety of rotational modes Set for full 360° rotation, or interrupt rotations with pausing & mixing intervals Slow rotating to vigorous mixing Cold room and incubator safe Accepts common tubes from 1.5 to 50mL Optional rocking platform Alkali Scientific™ RotoBot Mini R4040 includes holders for 12x1.5mL, 2x15mL and 2x50mL The RotoBot Rotating Mixers feature advanced programming options that far surpass that of the traditional laboratory rotator. With digital speed control and more than 10 operating modes, the RotoBot can be selected to perform a wide variety of rotating, mixing and vibrating actions. In addition, these actions can be combinesd in succession, using the C1 and C2 “Custom Modes”providing the user with a nearly infinite level of programming  
Catalog No. 50-302-9546 Supplier Alkali Scientific Supplier No. RS4040
Advanced programming with a wide variety of rotational modes Set for full 360° rotation, or interrupt rotations with pausing & mixing intervals Slow rotating to vigorous mixing Cold room and incubator safe Accepts common tubes from 1.5 to 50mL Optional rocking platform Alkali Scientific™ RotoBot Mini R4040 includes holders for 12x1.5mL, 2x15mL and 2x50mL The RotoBot Rotating Mixers feature advanced programming options that far surpass that of the traditional laboratory rotator. With digital speed control and more than 10 operating modes, the RotoBot can be selected to perform a wide variety of rotating, mixing and vibrating actions. In addition, these actions can be combinesd in succession, using the C1 and C2 “Custom Modes”providing the user with a nearly infinite level of programming  