Learn More
Sigma Aldrich Fine Chemicals Biosciences RotoBotTM Programmable Rot1EA
Catalog No. 502908761
Encompass
The RotoBot Rotating Mixers feature advanced programming options that far surpass that of the traditional laboratory rotator. With digital speed control and more than 10 operating modes the RotoBot can be selected to perform a wide variety of rotating mixing and vibrating actions. In addition these actions can be combined in succession using the C1 and C2 Custom Modes providing the user with a nearly infinite level of programming.The RotoBot and RotoBot Mini each inlcude an assortment of vertical tube holders. The Mini model is supplied with holders for 12 x 1.5 ml tubes 2 x 15 ml 2 x 50 ml tubes the standard model 30 x 1.5/2.0 ml 8 x 15 ml 2 x 50 ml tubes. For higher capacity applications more of these holders can be purchased as accessories. In addition horizontal holders are available for applications such as Western blotting and hybridization. The unique sliding attachment allows for quick and easy changing of tube holders.For rocking applications a platform with a no
Catalog No. 50-290-8761 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423001EA
The RotoBot Rotating Mixers feature advanced programming options that far surpass that of the traditional laboratory rotator. With digital speed control and more than 10 operating modes the RotoBot can be selected to perform a wide variety of rotating mixing and vibrating actions. In addition these actions can be combined in succession using the C1 and C2 Custom Modes providing the user with a nearly infinite level of programming.The RotoBot and RotoBot Mini each inlcude an assortment of vertical tube holders. The Mini model is supplied with holders for 12 x 1.5 ml tubes 2 x 15 ml 2 x 50 ml tubes the standard model 30 x 1.5/2.0 ml 8 x 15 ml 2 x 50 ml tubes. For higher capacity applications more of these holders can be purchased as accessories. In addition horizontal holders are available for applications such as Western blotting and hybridization. The unique sliding attachment allows for quick and easy changing of tube holders.For rocking applications a platform with a no