Learn More
Hettich Centrifuges ROTOFIX32A,(20)13s&(16)16s,Package
SDP
Catalog No. 502537626 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTOFIX 32A BLOOD TUBE PACKAGE BC, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7626 Supplier Hettich Centrifuges Supplier No. 32ABLOODBC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTOFIX 32A BLOOD TUBE PACKAGE BC, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years