Learn More
Hettich Centrifuges ROTOFIX32A,(20)Blood Tubes,Package
SDP
Catalog No. 502537627 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTOFIX 32A BLOOD TUBE PACKAGE, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7627 Supplier Hettich Centrifuges Supplier No. 32ABLOOD
Add to Cart
Add to Cart
Request a Quote
Hettich ROTOFIX 32A BLOOD TUBE PACKAGE, (1) Centrifuge, (1) Swing-out Rotor, (4) Buckets (4) Adapters 13 x 75/100, (4) Adapters 16 x 75/100, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years