Learn More
Cole-Parmer ROTOFLEX PLUS ROTATOR DIG DISP
Catalog No. NC0525516 Shop All Cole-Parmer Products
Encompass_Preferred
RotoFlex Plus Tube Rotator 120V is a variable speed, digitally controlled tube rotator; accommodating tubes from 0.5 - 50mL, digital display, dual operational mode (rotational, oscillating).
Catalog No. NC0525516 Supplier Cole-Parmer Supplier No. 0439740
RotoFlex Plus Tube Rotator 120V is a variable speed, digitally controlled tube rotator; accommodating tubes from 0.5 - 50mL, digital display, dual operational mode (rotational, oscillating).